Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Mdr49 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001169661.1 | NP_001163132.1 | 3 - 4 | 2162 | 66 | |
NM_079000.4 | NP_523724.2 | 6 - 7 | 2778 | 66 | ||
GenBank mRNA | BT044206.1 | - | 6 - 7 | 2756 | 66 | |
BT081353.1 | - | 3 - 4 | 2162 | 66 |
Set Membership: |
Amplicon length less than or equal to 70 Recommended Assay Drosophila melanogaster |