Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: tef | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_079042.4 | NP_523766.1 | 2 - 3 | 1540 | 88 | |
GenBank mRNA | AY051818.1 | - | 2 - 3 | 1515 | 88 |
Set Membership: |
Amplicon length between 86 and 100 3' Most Drosophila melanogaster |