Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Acox57D-p | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001299748.1 | NP_001286677.1 | 5 - 6 | 1674 | 84 | |
NM_079078.3 | NP_523802.1 | 5 - 6 | 1991 | 84 |
Set Membership: |
Amplicon length between 71 and 85 Drosophila melanogaster |