Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: gek | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_079113.3 | NP_523837.2 | 7 - 8 | 4444 | 86 | |
GenBank mRNA | AY051698.1 | - | 7 - 8 | 4425 | 86 |
Set Membership: |
Amplicon length between 86 and 100 Drosophila melanogaster |