Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Or63a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001169860.1 | NP_001163331.1 | 7 - 8 | 1335 | 59 | |
NM_079171.4 | NP_523895.2 | 8 - 9 | 1262 | 59 |
Set Membership: |
Amplicon length less than or equal to 70 G-protein coupled receptor Recommended Assay Drosophila melanogaster |