Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Prat2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_079225.3 | NP_523949.2 | 1 - 2 | 243 | 67 | |
GenBank mRNA | AF367369.1 | - | 1 - 2 | 229 | 67 | |
AY095010.1 | - | 1 - 2 | 222 | 67 |
Set Membership: |
Amplicon length less than or equal to 70 Drosophila melanogaster |