Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Or67a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_079281.4 | NP_524005.2 | 4 - 5 | 1388 | 86 | |
GenBank mRNA | BT032718.1 | - | 4 - 5 | 1388 | 86 |
Set Membership: |
Amplicon length between 86 and 100 G-protein coupled receptor 3' Most Drosophila melanogaster |