Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Nrx-IV | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_079310.3 | NP_524034.2 | 5 - 6 | 2075 | 81 | |
NM_168491.3 | NP_729787.2 | 5 - 6 | 2075 | 81 | ||
GenBank mRNA | BT120004.1 | - | 5 - 6 | 2042 | 81 |
Set Membership: |
Amplicon length between 71 and 85 Drosophila melanogaster |