Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: caup | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_079322.3 | NP_524046.2 | 2 - 3 | 1678 | 88 | |
GenBank mRNA | BT021246.1 | - | 2 - 3 | 1675 | 88 |
Set Membership: |
Amplicon length between 86 and 100 Drosophila melanogaster |