Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Prosbeta2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_079352.4 | NP_524076.2 | 2 - 3 | 359 | 58 | |
GenBank mRNA | AY051976.1 | - | 2 - 3 | 344 | 58 |
Set Membership: |
Amplicon length less than or equal to 70 3' Most Recommended Assay Drosophila melanogaster |