Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: mex1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001274951.1 | NP_001261880.1 | 1 - 2 | 475 | 53 | |
NM_079362.4 | NP_524086.1 | 1 - 2 | 151 | 53 | ||
GenBank mRNA | AY071615.1 | - | 1 - 2 | 147 | 53 | |
KX531514.1 | - | 1 - 2 | 50 | 53 |
Set Membership: |
Amplicon length less than or equal to 70 3' Most Recommended Assay Drosophila melanogaster |