Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Eig71Eg | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_079370.3 | NP_524094.2 | 2 - 3 | 263 | 101 |
Set Membership: |
Amplicon length greater than or equal to 101 3' Most Recommended Assay Drosophila melanogaster |