Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: comm | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001274962.1 | NP_001261891.1 | 1 - 2 | 733 | 83 | |
NM_079371.4 | NP_524095.1 | 1 - 2 | 733 | 83 | ||
GenBank mRNA | BT122139.1 | - | 1 - 2 | 725 | 83 | |
U35854.1 | - | 1 - 2 | 858 | 83 |
Set Membership: |
Amplicon length between 71 and 85 3' Most Recommended Assay Drosophila melanogaster |