Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Sin | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001170015.2 | NP_001163486.2 | 1 - 2 | 1218 | 61 | |
NM_079468.4 | NP_524192.2 | 2 - 3 | 1159 | 61 | ||
GenBank mRNA | BT126277.1 | - | 2 - 3 | 1127 | 61 |
Set Membership: |
Amplicon length less than or equal to 70 3' Most Recommended Assay Drosophila melanogaster |