Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Drl-2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001299442.1 | NP_001286371.1 | 10 - 11 | 3046 | 77 | |
NM_136991.4 | NP_610835.2 | 10 - 11 | 3046 | 77 | ||
GenBank mRNA | BT004493.1 | - | 10 - 11 | 3017 | 77 |
Set Membership: |
Amplicon length between 71 and 85 3' Most Drosophila melanogaster |