Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: spirit | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001169236.2 | NP_001162707.1 | 2 - 3 | 197 | 89 | |
NM_001272413.1 | NP_001259342.1 | 2 - 3 | 246 | 89 | ||
NM_132264.3 | NP_572492.1 | 1 - 2 | 226 | 89 | ||
NM_167161.2 | NP_727276.1 | 2 - 3 | 230 | 89 | ||
GenBank mRNA | AY058283.1 | - | 1 - 2 | 226 | 89 |
Set Membership: |
Amplicon length between 86 and 100 Drosophila melanogaster |