Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ppt1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001298095.2 | NP_001285024.1 | 4 - 5 | 1012 | 66 | |
NM_167166.4 | NP_727284.1 | 3 - 4 | 891 | 66 | ||
GenBank mRNA | AF513720.1 | - | 3 - 4 | 833 | 66 | |
BT150202.1 | - | 3 - 4 | 889 | 66 |
Set Membership: |
Amplicon length less than or equal to 70 3' Most Recommended Assay Drosophila melanogaster |