Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ir8a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_167185.1 | NP_727328.1 | 3 - 4 | 604 | 61 |
Set Membership: |
Amplicon length less than or equal to 70 Recommended Assay Drosophila melanogaster |