Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Gr9a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_167217.1 | NP_727392.1 | 2 - 3 | 953 | 135 |
Set Membership: |
Amplicon length greater than or equal to 101 3' Most Drosophila melanogaster |