Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: X11Lbeta | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001272496.2 | NP_001259425.2 | 7 - 8 | 5547 | 96 | |
NM_001298156.1 | NP_001285085.1 | 9 - 10 | 4674 | 96 | ||
NM_167239.4 | NP_727440.3 | 9 - 10 | 4629 | 96 |
Set Membership: |
Amplicon length between 86 and 100 Drosophila melanogaster |