Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: eIF4G1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001103382.2 | NP_001096852.1 | 15 - 16 | 5977 | 74 | |
NM_001144385.2 | NP_001137857.1 | 14 - 15 | 5448 | 74 | ||
NM_079901.4 | NP_524640.3 | 14 - 15 | 5218 | 74 | ||
GenBank mRNA | AF030155.1 | - | 14 - 15 | 5224 | 74 | |
BT053671.1 | - | 14 - 15 | 5212 | 74 |
Set Membership: |
Amplicon length between 71 and 85 Drosophila melanogaster |