Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Shc | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_079944.3 | NP_524683.2 | 1 - 2 | 310 | 72 | |
GenBank mRNA | BT044327.1 | - | 1 - 2 | 319 | 72 |
Set Membership: |
Amplicon length between 71 and 85 3' Most Recommended Assay Drosophila melanogaster |