Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: RpL8 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_079987.3 | NP_524726.1 | 2 - 3 | 195 | 59 | |
NM_167955.2 | NP_728756.1 | 2 - 3 | 370 | 59 | ||
GenBank mRNA | AY071342.1 | - | 2 - 3 | 199 | 59 | |
BT014913.1 | - | 1626 | 59 |
Set Membership: |
Amplicon length less than or equal to 70 3' Most Recommended Assay Drosophila melanogaster Inventoried |