Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: icln | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_137393.4 | NP_611237.2 | 1 - 2 | 190 | 66 |
Set Membership: |
Amplicon length less than or equal to 70 3' Most Recommended Assay Drosophila melanogaster Inventoried |