Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Sec6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_137507.5 | NP_611351.1 | 1 - 2 | 1061 | 80 | |
GenBank mRNA | AY058549.1 | - | 1 - 2 | 1032 | 80 |
Set Membership: |
Amplicon length between 71 and 85 3' Most Recommended Assay Drosophila melanogaster |