Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Tao | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001201743.3 | NP_001188672.1 | 3 - 4 | 1232 | 85 | |
NM_001258827.1 | NP_001245756.1 | 3 - 4 | 1172 | 85 | ||
NM_001272780.2 | NP_001259709.1 | 3 - 4 | 1232 | 85 | ||
NM_167665.2 | NP_728267.1 | 3 - 4 | 882 | 85 | ||
NM_167666.3 | NP_728268.1 | 3 - 4 | 1232 | 85 | ||
GenBank mRNA | AB277547.1 | - | 3 - 4 | 1211 | 85 | |
BT099719.1 | - | 3 - 4 | 1171 | 85 |
Set Membership: |
Amplicon length between 71 and 85 Drosophila melanogaster |