Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: tw | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001272173.1 | NP_001259102.1 | 6 - 7 | 1438 | 85 | |
NM_130502.3 | NP_569858.1 | 6 - 7 | 1438 | 85 | ||
GenBank mRNA | AB176551.1 | - | 6 - 7 | 1279 | 85 | |
AY095525.1 | - | 6 - 7 | 1393 | 85 |
Set Membership: |
Amplicon length between 71 and 85 Drosophila melanogaster |