Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Gmer | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_137890.3 | NP_611734.1 | 2 - 3 | 340 | 82 | |
GenBank mRNA | AM231688.1 | - | 2 - 3 | 242 | 82 | |
AY060873.1 | - | 2 - 3 | 275 | 82 | ||
BT050562.1 | - | 2 - 3 | 335 | 82 |
Set Membership: |
Amplicon length between 71 and 85 Drosophila melanogaster |