Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: levy | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001299841.1 | NP_001286770.1 | 1 - 2 | 147 | 94 | |
NM_137961.3 | NP_611805.1 | 1 - 2 | 147 | 94 | ||
GenBank mRNA | AY070648.1 | - | 1 - 2 | 146 | 94 |
Set Membership: |
Amplicon length between 86 and 100 3' Most Recommended Assay Drosophila melanogaster |