Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: shu | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_137993.4 | NP_611837.1 | 1 - 2 | 151 | 67 | |
GenBank mRNA | AY058554.1 | - | 1 - 2 | 121 | 67 |
Set Membership: |
Amplicon length less than or equal to 70 Recommended Assay Drosophila melanogaster |