Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Zfrp8 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001299899.1 | NP_001286828.1 | 2 - 3 | 385 | 58 | |
NM_138046.2 | NP_611890.1 | 2 - 3 | 385 | 58 |
Set Membership: |
Amplicon length less than or equal to 70 3' Most Recommended Assay Drosophila melanogaster |