Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ITP | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001043104.3 | NP_001036569.2 | 1 - 2 | 1080 | 72 | |
NM_001169822.3 | NP_001163293.1 | 1 - 2 | 1080 | 72 | ||
NM_138087.6 | NP_611931.3 | 1 - 2 | 1080 | 72 | ||
GenBank mRNA | AY060468.1 | - | 1 - 2 | 1056 | 72 |
Set Membership: |
Amplicon length between 71 and 85 3' Most Drosophila melanogaster |