Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Spt6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001298007.1 | NP_001284936.1 | 5 - 6 | 715 | 67 | |
NM_143705.3 | NP_651962.2 | 5 - 6 | 715 | 67 | ||
GenBank mRNA | BT150092.1 | - | 5 - 6 | 689 | 67 | |
BT150473.1 | - | 5 - 6 | 689 | 67 |
Set Membership: |
Amplicon length less than or equal to 70 3' Most Drosophila melanogaster |