Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Rs1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001299259.1 | NP_001286188.1 | 2 - 3 | 304 | 78 | |
NM_143713.3 | NP_651970.1 | 1 - 2 | 161 | 78 | ||
GenBank mRNA | BT004908.1 | - | 1 - 2 | 121 | 78 |
Set Membership: |
Amplicon length between 71 and 85 Drosophila melanogaster |