Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: mab-21 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_143714.4 | NP_651971.2 | 1 - 2 | 1107 | 72 | |
GenBank mRNA | AY128466.1 | - | 1 - 2 | 1104 | 72 |
Set Membership: |
Amplicon length between 71 and 85 3' Most Recommended Assay Drosophila melanogaster |