Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ady43A | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_143738.4 | NP_651995.1 | 2 - 3 | 706 | 76 | |
GenBank mRNA | AY102669.1 | - | 2 - 3 | 724 | 76 |
Set Membership: |
Amplicon length between 71 and 85 3' Most Recommended Assay Drosophila melanogaster |