Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Prosbeta1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_143774.3 | NP_652031.2 | 2 - 3 | 613 | 64 | |
GenBank mRNA | BT044319.1 | - | 2 - 3 | 607 | 64 |
Set Membership: |
Amplicon length less than or equal to 70 3' Most Recommended Assay Drosophila melanogaster |