Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Fis1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001144135.2 | NP_001137607.1 | 5 - 6 | 530 | 109 | |
NM_001299211.1 | NP_001286140.1 | 3 - 4 | 389 | 109 | ||
NM_143843.4 | NP_652100.1 | 4 - 5 | 462 | 109 | ||
GenBank mRNA | AY071264.1 | - | 4 - 5 | 456 | 109 |
Set Membership: |
Amplicon length greater than or equal to 101 3' Most Drosophila melanogaster |