Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Iml1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001274330.1 | NP_001261259.1 | 16 - 17 | 3827 | 62 | |
NM_001274331.1 | NP_001261260.1 | 16 - 17 | 3803 | 62 | ||
NM_139361.2 | NP_647618.1 | 15 - 16 | 3707 | 62 | ||
NM_167888.2 | NP_728620.1 | 15 - 16 | 3710 | 62 | ||
NM_167889.2 | NP_728621.1 | 17 - 18 | 3926 | 62 |
Set Membership: |
Amplicon length less than or equal to 70 Drosophila melanogaster |