Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: dre4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_057262.4 | NP_476610.2 | 2 - 3 | 465 | 76 | |
NM_167922.3 | NP_728686.2 | 2 - 3 | 465 | 76 | ||
GenBank mRNA | BT010116.1 | - | 2 - 3 | 489 | 76 |
Set Membership: |
Amplicon length between 71 and 85 Drosophila melanogaster |