Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: shep | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_168112.4 | NP_729055.1 | 9 - 10 | 1993 | 112 | |
NM_168113.3 | NP_729056.1 | 8 - 9 | 3444 | 112 |
Set Membership: |
Amplicon length greater than or equal to 101 3' Most Drosophila melanogaster |