Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Uev1A | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001300011.1 | NP_001286940.1 | 3 - 4 | 321 | 91 | |
NM_139702.4 | NP_647959.1 | 3 - 4 | 364 | 91 | ||
NM_168117.3 | NP_729062.1 | 3 - 4 | 475 | 91 | ||
GenBank mRNA | AY060384.1 | - | 3 - 4 | 300 | 91 | |
BT120202.1 | - | 3 - 4 | 476 | 91 | ||
KX531865.1 | - | 3 - 4 | 295 | 91 | ||
KX671622.1 | - | 2 - 3 | 169 | 91 |
Set Membership: |
Amplicon length between 86 and 100 3' Most Drosophila melanogaster |