Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Or65c | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_168165.2 | NP_729163.2 | 2 - 3 | 286 | 92 |
Set Membership: |
Amplicon length between 86 and 100 G-protein coupled receptor Drosophila melanogaster |