Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: HP4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_168247.3 | NP_729314.1 | 1 | 473 | 113 | |
GenBank mRNA | BT132888.1 | - | 1 | 431 | 113 |
Set Membership: |
Amplicon length greater than or equal to 101 3' Most Drosophila melanogaster |