Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Sarm | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001043129.3 | NP_001036594.1 | 4 - 5 | 3350 | 66 | |
NM_001104071.2 | NP_001097541.1 | 5 - 6 | 3498 | 66 | ||
NM_001104074.3 | NP_001097544.1 | 6 - 7 | 3432 | 66 | ||
NM_001104075.2 | NP_001097545.1 | 5 - 6 | 3256 | 66 | ||
NM_001259733.1 | NP_001246662.1 | 7 - 8 | 5043 | 66 | ||
NM_001259734.1 | NP_001246663.1 | 4 - 5 | 3050 | 66 | ||
NM_001274627.1 | NP_001261556.1 | 6 - 7 | 4456 | 66 | ||
NM_001382096.1 | NP_001369041.1 | 2 - 3 | 2242 | 66 | ||
NM_168254.2 | NP_729327.1 | 3 - 4 | 2851 | 66 | ||
GenBank mRNA | AY075346.1 | - | 3 - 4 | 2832 | 66 | |
BT014666.1 | - | 5 - 6 | 3230 | 66 |
Set Membership: |
Amplicon length less than or equal to 70 Recommended Assay Drosophila melanogaster |