Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Vha16-2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_168459.2 | NP_729707.1 | 1 | 399 | 70 | |
GenBank mRNA | KX531991.1 | - | 1 | 359 | 70 |
Set Membership: |
Amplicon length less than or equal to 70 3' Most Recommended Assay Drosophila melanogaster |