Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Best4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001104139.2 | NP_001097609.1 | 4 - 5 | 1159 | 73 |
Set Membership: |
Amplicon length between 71 and 85 Drosophila melanogaster |