Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Mbs | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001104144.3 | NP_001097614.1 | 13 - 14 | 2563 | 59 | |
NM_001104145.3 | NP_001097615.1 | 14 - 15 | 2946 | 59 | ||
NM_001259854.2 | NP_001246783.1 | 15 - 16 | 4031 | 59 | ||
NM_001259855.2 | NP_001246784.1 | 16 - 17 | 3931 | 59 | ||
NM_001259856.2 | NP_001246785.1 | 16 - 17 | 4187 | 59 | ||
NM_001259857.2 | NP_001246786.1 | 16 - 17 | 3934 | 59 | ||
NM_001259858.2 | NP_001246787.1 | 14 - 15 | 3002 | 59 | ||
NM_001274990.1 | NP_001261919.1 | 14 - 15 | 3394 | 59 | ||
NM_001382059.1 | NP_001369057.1 | 14 - 15 | 3394 | 59 | ||
NM_168646.3 | NP_730099.2 | 15 - 16 | 3516 | 59 | ||
NM_168647.3 | NP_730100.2 | 14 - 15 | 2758 | 59 | ||
NM_168648.3 | NP_730101.2 | 14 - 15 | 3369 | 59 | ||
GenBank mRNA | AF500094.1 | - | 13 - 14 | 2901 | 59 | |
BT133053.1 | - | 15 - 16 | 3504 | 59 |
Set Membership: |
Amplicon length less than or equal to 70 Drosophila melanogaster |