Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Cyp4e1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_080032.3 | NP_524771.1 | 4 - 5 | 1039 | 83 | |
GenBank mRNA | AY118793.1 | - | 4 - 5 | 1020 | 83 |
Set Membership: |
Amplicon length between 71 and 85 Drosophila melanogaster |