Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Hmr | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_132409.3 | NP_572637.2 | 2 - 3 | 3078 | 69 |
Set Membership: |
Amplicon length less than or equal to 70 Drosophila melanogaster |